Assay Details
| Gene Symbol: KRT13 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001001312.2 | NP_001001312.2 | 5 - 6 | 1118 | 68 | |
| GenBank mRNA | AY574986.1 | - | 6 - 7 | 1020 | 68 | |
| BU389884.1 | - | 1 - 2 | 53 | 68 | ||
| CN227144.1 | - | 3 - 4 | 380 | 68 | ||
More Information
Set Membership: |
|

KRT13
Genomic Map