Assay Details
| Gene Symbol: ST8SIA5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001001747.2 | NP_001001747.2 | 9 - 10 | 1638 | 73 | |
| XM_004949029.3 | XP_004949086.1 | 10 - 11 | 1676 | 73 | ||
| XM_004949031.3 | XP_004949088.1 | 8 - 9 | 4991 | 73 | ||
| XM_004949032.3 | XP_004949089.1 | 8 - 9 | 1474 | 73 | ||
| XM_015277412.2 | XP_015132898.1 | 9 - 10 | 5083 | 73 | ||
| XM_015277413.2 | XP_015132899.1 | 8 - 9 | 1428 | 73 | ||
| XM_015277414.2 | XP_015132900.1 | 8 - 9 | 1446 | 73 | ||
| XM_015277415.2 | XP_015132901.1 | 7 - 8 | 4883 | 73 | ||
| XM_015277416.1 | XP_015132902.1 | 7 - 8 | 664 | 73 | ||
| GenBank mRNA | AJ704564.1 | - | 5 - 6 | 664 | 73 | |
More Information
Set Membership: |
|

ST8SIA5
Genomic Map