Assay Details
| Gene Symbol: EPYC | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001004382.2 | NP_001004382.1 | 5 - 6 | 726 | 97 | |
| XM_025148153.1 | XP_025003921.1 | 5 - 6 | 749 | 97 | ||
| GenBank mRNA | D10485.1 | - | 5 - 6 | 726 | 97 | |
More Information
Set Membership: |
|

EPYC
Genomic Map