Assay Details
| Gene Symbol: ZYX | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001004386.1 | NP_001004386.1 | 4 - 5 | 947 | 81 | |
| XM_015293412.1 | XP_015148898.1 | 5 - 6 | 1195 | 81 | ||
| XM_015293419.2 | XP_015148905.1 | 4 - 5 | 1102 | 81 | ||
| XM_015293430.2 | XP_015148916.1 | 5 - 6 | 1034 | 81 | ||
| XM_015293437.2 | XP_015148923.1 | 5 - 6 | 1153 | 81 | ||
| XM_015293439.2 | XP_015148925.1 | 5 - 6 | 1164 | 81 | ||
| XM_015293446.2 | XP_015148932.1 | 5 - 6 | 1143 | 81 | ||
| XM_025149192.1 | XP_025004960.1 | 4 - 5 | 1020 | 81 | ||
| GenBank mRNA | X69190.1 | - | 947 | 81 | ||
More Information
Set Membership: |
|

ZYX
Genomic Map