Assay Details
| Gene Symbol: VDHAP | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_204976.2 | NP_990307.2 | 10 - 11 | 1421 | 74 | |
| GenBank mRNA | BU223174.1 | - | 2 - 3 | 181 | 74 | |
| U00694.1 | - | 10 - 11 | 1454 | 74 | ||
More Information
Set Membership: |
|

VDHAP
Genomic Map