Assay Details
| Gene Symbol: VDHAP | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_204976.2 | NP_990307.2 | 11 - 12 | 1464 | 103 | |
| GenBank mRNA | BU223174.1 | - | 3 - 4 | 224 | 103 | |
| U00694.1 | - | 11 - 12 | 1497 | 101 | ||
More Information
Set Membership: |
|

VDHAP
Genomic Map