Assay Details
| Gene Symbol: VDHAP | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_204976.2 | NP_990307.2 | 1 - 2 | 332 | 60 | |
| GenBank mRNA | U00694.1 | - | 1 - 2 | 365 | 60 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

VDHAP| Gene Symbol: VDHAP | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_204976.2 | NP_990307.2 | 1 - 2 | 332 | 60 | |
| GenBank mRNA | U00694.1 | - | 1 - 2 | 365 | 60 | |
Set Membership: |
|