Assay Details
Gene Symbol: GCM1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_206980.1 | NP_996863.1 | 4 - 5 | 584 | 66 | |
XM_015284823.2 | XP_015140309.1 | 4 - 5 | 632 | 66 | ||
GenBank mRNA | AY371201.1 | - | 4 - 5 | 584 | 66 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
