Assay Details
| Gene Symbol: TNMD | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_206985.2 | NP_996868.1 | 4 - 5 | 465 | 66 | |
| GenBank mRNA | AY156693.1 | - | 4 - 5 | 467 | 66 | |
| BX935041.1 | - | 3 - 4 | 620 | 66 | ||
More Information
Set Membership: |
|

TNMD
Genomic Map