Assay Details
| Gene Symbol: MATN1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001030375.2 | NP_001025546.2 | 3 - 4 | 692 | 61 | |
| GenBank mRNA | M14792.1 | - | 2 - 3 | 423 | 61 | |
More Information
Set Membership: |
|

MATN1
Genomic Map