Assay Details
| Gene Symbol: C26H6orf106 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001030919.1 | NP_001026090.1 | 220 | 92 | ||
| GenBank mRNA | AJ851611.1 | - | 220 | 92 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

C26H6orf106| Gene Symbol: C26H6orf106 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001030919.1 | NP_001026090.1 | 220 | 92 | ||
| GenBank mRNA | AJ851611.1 | - | 220 | 92 | ||
Set Membership: |
|