Assay Details
| Gene Symbol: C26H6orf106 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001030919.1 | NP_001026090.1 | 569 | 79 | ||
| XM_015298936.1 | XP_015154422.1 | 434 | 79 | |||
| GenBank mRNA | AJ851611.1 | - | 569 | 79 | ||
More Information
Set Membership: |
|

C26H6orf106
Genomic Map