Assay Details
Gene Symbol: RPL19 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001030929.1 | NP_001026100.1 | 5 - 6 | 502 | 71 | |
GenBank mRNA | AJ720076.1 | - | 5 - 6 | 502 | 71 | |
AY393838.1 | - | 4 - 5 | 447 | 71 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
