Assay Details
| Gene Symbol: DCTN3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001031445.1 | NP_001026616.1 | 3 - 4 | 291 | 63 | |
| GenBank mRNA | AJ720265.1 | - | 3 - 4 | 291 | 63 | |
| BX932189.2 | - | 3 - 4 | 271 | 63 | ||
More Information
Set Membership: |
|

DCTN3
Genomic Map