Assay Details
| Gene Symbol: AGRP | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001031457.1 | NP_001026628.1 | 2 - 3 | 301 | 75 | |
| XM_025154207.1 | XP_025009975.1 | 3 - 4 | 400 | 75 | ||
| GenBank mRNA | AB489991.1 | - | 3 - 4 | 342 | 75 | |
| AB489992.1 | - | 3 - 4 | 425 | 75 | ||
| AB489993.1 | - | 2 - 3 | 759 | 75 | ||
More Information
Set Membership: |
|

AGRP
Genomic Map