Assay Details
| Gene Symbol: SLC41A2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001031483.1 | NP_001026654.1 | 8 - 9 | 1523 | 72 | |
| XM_015286032.2 | XP_015141518.1 | 7 - 8 | 1507 | 72 | ||
| XM_015286035.2 | XP_015141521.1 | 8 - 9 | 1763 | 72 | ||
| GenBank mRNA | AJ721008.1 | - | 8 - 9 | 1523 | 72 | |
More Information
Set Membership: |
|

SLC41A2
Genomic Map