Assay Details
| Gene Symbol: CHMP6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001031529.1 | NP_001026700.1 | 2 - 3 | 229 | 66 | |
| GenBank mRNA | AJ719879.1 | - | 2 - 3 | 229 | 66 | |
More Information
Set Membership: |
|

CHMP6
Genomic Map