Assay Details
| Gene Symbol: SET | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001030691.2 | NP_001025862.2 | 1 - 2 | 351 | 69 | |
| GenBank mRNA | AJ851581.1 | - | 1 - 2 | 88 | 69 | |
More Information
Set Membership: |
|

SET
Genomic Map