Assay Details
Gene Symbol: HNRNPA2B1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001030985.1 | NP_001026156.1 | 2 - 3 | 292 | 93 | |
XM_015281456.2 | XP_015136942.1 | 2 - 3 | 298 | 93 | ||
XM_015281458.2 | XP_015136944.1 | 2 - 3 | 298 | 93 | ||
XM_015281462.2 | XP_015136948.1 | 2 - 3 | 299 | 93 | ||
XM_025147198.1 | XP_025002966.1 | 2 - 3 | 272 | 93 | ||
XM_025147199.1 | XP_025002967.1 | 2 - 3 | 299 | 93 | ||
XM_025147200.1 | XP_025002968.1 | 2 - 3 | 387 | 93 | ||
XM_025147201.1 | XP_025002969.1 | 2 - 3 | 455 | 93 | ||
XM_025147202.1 | XP_025002970.1 | 2 - 3 | 298 | 93 | ||
XM_025147203.1 | XP_025002971.1 | 2 - 3 | 298 | 93 | ||
GenBank mRNA | AJ719443.1 | - | 2 - 3 | 292 | 93 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
