Assay Details
| Gene Symbol: DAP | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001031003.1 | NP_001026174.1 | 1 - 2 | 94 | 82 | |
| GenBank mRNA | AJ720932.1 | - | 1 - 2 | 94 | 82 | |
| JQ268269.1 | - | 1 - 2 | 63 | 82 | ||
More Information
Set Membership: |
|

DAP
Genomic Map