Assay Details
| Gene Symbol: THAP10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001031277.1 | NP_001026448.1 | 1 - 2 | 377 | 69 | |
| XM_015290819.2 | XP_015146305.1 | 1 - 2 | 255 | 69 | ||
| GenBank mRNA | AJ851595.1 | - | 1 - 2 | 377 | 69 | |
More Information
Set Membership: |
|

THAP10
Genomic Map