Assay Details
| Gene Symbol: MVB12A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001031355.1 | NP_001026526.1 | 5 - 6 | 541 | 65 | |
| GenBank mRNA | AJ720307.1 | - | 5 - 6 | 541 | 65 | |
| BX931276.1 | - | 5 - 6 | 527 | 65 | ||
| BX931683.2 | - | 5 - 6 | 516 | 65 | ||
| CR354124.1 | - | 5 - 6 | 542 | 65 | ||
More Information
Set Membership: |
|

MVB12A
Genomic Map