Assay Details
| Gene Symbol: TNFRSF13C | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001037828.1 | NP_001032917.1 | 2 - 3 | 341 | 134 | |
| XM_025148512.1 | XP_025004280.1 | 2 - 3 | 1066 | 134 | ||
| GenBank mRNA | AM085437.1 | - | 2 - 3 | 341 | 134 | |
More Information
Set Membership: |
|

TNFRSF13C
Genomic Map