Assay Details
| Gene Symbol: ZDHHC8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001038691.1 | NP_001033780.1 | 8 - 9 | 1034 | 59 | |
| XM_015275255.2 | XP_015130741.1 | 7 - 8 | 3617 | 59 | ||
| XM_015275256.2 | XP_015130742.1 | 8 - 9 | 1157 | 59 | ||
| XM_015275257.2 | XP_015130743.1 | 7 - 8 | 848 | 59 | ||
| GenBank mRNA | AY871207.1 | - | 8 - 9 | 1034 | 59 | |
| BX930153.1 | - | 5 - 6 | 501 | 59 | ||
More Information
Set Membership: |
|

ZDHHC8
Genomic Map