Assay Details
| Gene Symbol: ACAT2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001039287.2 | NP_001034376.2 | 3 - 4 | 425 | 56 | |
| GenBank mRNA | AJ721080.1 | - | 3 - 4 | 426 | 56 | |
More Information
Set Membership: |
|

ACAT2
Genomic Map