Assay Details
| Gene Symbol: VAMP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001039486.1 | NP_001034575.1 | 2 - 3 | 150 | 91 | |
| XM_015293316.2 | XP_015148802.1 | 2 - 3 | 225 | 91 | ||
| XM_025149125.1 | XP_025004893.1 | 2 - 3 | 225 | 91 | ||
| XM_025149131.1 | XP_025004899.1 | 2 - 3 | 225 | 91 | ||
| GenBank mRNA | BM490145.1 | - | 2 - 3 | 150 | 91 | |
More Information
Set Membership: |
|

VAMP1
Genomic Map