Assay Details
| Gene Symbol: KLHL18 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001030960.2 | NP_001026131.1 | 9 - 10 | 1498 | 53 | |
| XM_015281088.2 | XP_015136574.1 | 9 - 10 | 1481 | 53 | ||
| XM_015281089.2 | XP_015136575.1 | 9 - 10 | 1370 | 53 | ||
| XM_015281090.2 | XP_015136576.2 | 9 - 10 | 1408 | 53 | ||
| XM_015281091.2 | XP_015136577.1 | 9 - 10 | 1443 | 53 | ||
| XM_015281092.2 | XP_015136578.1 | 9 - 10 | 1383 | 53 | ||
| XM_015281093.2 | XP_015136579.1 | 9 - 10 | 1437 | 53 | ||
| XM_025147151.1 | XP_025002919.1 | 9 - 10 | 1461 | 53 | ||
| XM_025147152.1 | XP_025002920.1 | 9 - 10 | 1360 | 53 | ||
| GenBank mRNA | AJ720960.1 | - | 9 - 10 | 1498 | 53 | |
More Information
Set Membership: |
|

KLHL18
Genomic Map