Assay Details
| Gene Symbol: RASD1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001044636.1 | NP_001038101.1 | 1 - 2 | 365 | 59 | |
| GenBank mRNA | DQ516976.1 | - | 1 - 2 | 365 | 59 | |
More Information
Set Membership: |
|

RASD1
Genomic Map