Assay Details
| Gene Symbol: ASF1A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001044690.1 | NP_001038155.1 | 3 - 4 | 667 | 94 | |
| GenBank mRNA | AB238225.1 | - | 3 - 4 | 667 | 94 | |
More Information
Set Membership: |
|

ASF1A
Genomic Map