Assay Details
| Gene Symbol: PCDH8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001098609.1 | NP_001092079.1 | 2 - 3 | 2623 | 73 | |
| XM_015275767.2 | XP_015131253.1 | 2 - 3 | 2722 | 73 | ||
| GenBank mRNA | EF175382.1 | - | 2 - 3 | 2623 | 73 | |
| JN252709.1 | - | 2 - 3 | 2482 | 73 | ||
More Information
Set Membership: |
|

PCDH8
Genomic Map