Assay Details
| Gene Symbol: BAG1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001109692.1 | NP_001103162.1 | 5 - 6 | 525 | 98 | |
| XM_015282328.2 | XP_015137814.1 | 4 - 5 | 395 | 98 | ||
| GenBank mRNA | CR386184.1 | - | 5 - 6 | 496 | 98 | |
More Information
Set Membership: |
|

BAG1
Genomic Map