Assay Details
Gene Symbol: CEBPB | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_205253.2 | NP_990584.1 | 1 | 647 | 116 | |
GenBank mRNA | X70813.1 | - | 436 | 116 | ||
Z21646.1 | - | 647 | 116 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
