Assay Details
| Gene Symbol: LPAR1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001115082.1 | NP_001108554.1 | 2 - 3 | 808 | 89 | |
| XM_015280609.2 | XP_015136095.1 | 3 - 4 | 1000 | 89 | ||
| XM_015280610.2 | XP_015136096.1 | 3 - 4 | 1099 | 89 | ||
| GenBank mRNA | BX930927.2 | - | 2 - 3 | 852 | 89 | |
| BX935575.1 | - | 3 - 4 | 969 | 89 | ||
| EU339317.1 | - | 2 - 3 | 808 | 89 | ||
More Information
Set Membership: |
|

LPAR1
Genomic Map