Assay Details
| Gene Symbol: PAFAH1B1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_204324.1 | NP_989655.1 | 6 - 7 | 799 | 64 | |
| XM_015295659.2 | XP_015151145.1 | 6 - 7 | 780 | 64 | ||
| XM_015295660.1 | XP_015151146.1 | 7 - 8 | 800 | 64 | ||
| XM_015295661.2 | XP_015151147.1 | 7 - 8 | 968 | 64 | ||
| XM_025141880.1 | XP_024997648.1 | 8 - 9 | 7679 | 64 | ||
| XM_025141881.1 | XP_024997649.1 | 8 - 9 | 7704 | 64 | ||
| GenBank mRNA | AF113946.1 | - | 6 - 7 | 799 | 64 | |
More Information
Set Membership: |
|

PAFAH1B1
Genomic Map