Assay Details
| Gene Symbol: LIX1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_204347.1 | NP_989678.1 | 2 - 3 | 242 | 59 | |
| GenBank mRNA | AF351203.1 | - | 2 - 3 | 242 | 59 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

LIX1| Gene Symbol: LIX1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_204347.1 | NP_989678.1 | 2 - 3 | 242 | 59 | |
| GenBank mRNA | AF351203.1 | - | 2 - 3 | 242 | 59 | |
Set Membership: |
|