Assay Details
Gene Symbol: ST6GALNAC4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_204511.1 | NP_989842.1 | 2 - 3 | 220 | 108 | |
XM_015279303.2 | XP_015134789.2 | 3 - 4 | 536 | 108 | ||
XM_015279304.2 | XP_015134790.2 | 3 - 4 | 512 | 108 | ||
GenBank mRNA | AJ620653.1 | - | 2 - 3 | 220 | 108 | |
BX929951.1 | - | 2 - 3 | 221 | 108 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
