Assay Details
| Gene Symbol: SOCS2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_204540.1 | NP_989871.1 | 1 | 450 | 64 | |
| GenBank mRNA | AF498236.1 | - | 1 | 446 | 64 | |
| AF540005.1 | - | 2 | 360 | 64 | ||
More Information
Set Membership: |
|

SOCS2
Genomic Map