Assay Details
| Gene Symbol: FSHR | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_205079.1 | NP_990410.1 | 8 - 9 | 723 | 76 | |
| XM_025148544.1 | XP_025004312.1 | 8 - 9 | 988 | 76 | ||
| GenBank mRNA | D87871.1 | - | 8 - 9 | 723 | 76 | |
| U51097.1 | - | 8 - 9 | 691 | 76 | ||
More Information
Set Membership: |
|

FSHR
Genomic Map