Assay Details
| Gene Symbol: FTH1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_205086.1 | NP_990417.1 | 1 - 2 | 261 | 61 | |
| GenBank mRNA | BX934496.1 | - | 1 - 2 | 255 | 61 | |
| BX934547.1 | - | 1 - 2 | 232 | 61 | ||
| Y14698.1 | - | 261 | 61 | |||
More Information
Set Membership: |
|

FTH1
Genomic Map