Assay Details
| Gene Symbol: GFRA2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_205101.1 | NP_990432.1 | 6 - 7 | 1077 | 58 | |
| GenBank mRNA | U90542.1 | - | 6 - 7 | 1077 | 58 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

GFRA2| Gene Symbol: GFRA2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_205101.1 | NP_990432.1 | 6 - 7 | 1077 | 58 | |
| GenBank mRNA | U90542.1 | - | 6 - 7 | 1077 | 58 | |
Set Membership: |
|