Assay Details
| Gene Symbol: CYP1A1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_205147.1 | NP_990478.1 | 2 - 3 | 929 | 72 | |
| GenBank mRNA | X99453.1 | - | 929 | 72 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

CYP1A1| Gene Symbol: CYP1A1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_205147.1 | NP_990478.1 | 2 - 3 | 929 | 72 | |
| GenBank mRNA | X99453.1 | - | 929 | 72 | ||
Set Membership: |
|