Assay Details
| Gene Symbol: CCZ1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001006158.1 | NP_001006158.1 | 4 - 5 | 470 | 64 | |
| GenBank mRNA | AJ719702.1 | - | 4 - 5 | 470 | 64 | |
More Information
Set Membership: |
|

CCZ1
Genomic Map