Assay Details
Gene Symbol: POC1B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001006232.1 | NP_001006232.1 | 11 - 12 | 1397 | 69 | |
GenBank mRNA | AJ719544.1 | - | 11 - 12 | 1397 | 69 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
