Assay Details
| Gene Symbol: TMEM263 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001006244.1 | NP_001006244.1 | 4 - 5 | 320 | 61 | |
| XM_015285437.2 | XP_015140923.1 | 4 - 5 | 617 | 61 | ||
| XM_025148704.1 | XP_025004472.1 | 5 - 6 | 532 | 61 | ||
| GenBank mRNA | AJ719825.1 | - | 4 - 5 | 320 | 61 | |
More Information
Set Membership: |
|

TMEM263
Genomic Map