Assay Details
Gene Symbol: ICMT | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001006298.1 | NP_001006298.1 | 4 - 5 | 734 | 62 | |
GenBank mRNA | AJ719287.1 | - | 4 - 5 | 734 | 62 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Panther Classification:
