Assay Details
| Gene Symbol: HSPA2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001006685.1 | NP_001006686.1 | 1 | 1844 | 102 | |
| GenBank mRNA | FJ217667.1 | - | 1 | 1747 | 102 | |
More Information
Set Membership: |
|

HSPA2
Genomic Map