Assay Details
| Gene Symbol: TMED5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001007956.1 | NP_001007957.1 | 1 - 2 | 225 | 146 | |
| GenBank mRNA | AJ719300.1 | - | 1 - 2 | 225 | 146 | |
| CR389193.1 | - | 1 - 2 | 204 | 146 | ||
More Information
Set Membership: |
|

TMED5
Genomic Map