Assay Details
| Gene Symbol: DCLRE1A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001008683.1 | NP_001008683.1 | 4 - 5 | 2165 | 84 | |
| XM_015288657.1 | XP_015144143.1 | 4 - 5 | 2700 | 84 | ||
| GenBank mRNA | AY376896.1 | - | 4 - 5 | 2165 | 84 | |
More Information
Set Membership: |
|

DCLRE1A
Genomic Map