Assay Details
Gene Symbol: CASP6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_204726.1 | NP_990057.1 | 1 - 2 | 48 | 68 | |
XM_015276125.2 | XP_015131611.1 | 1 - 2 | 4450 | 68 | ||
XM_025149501.1 | XP_025005269.1 | 1 - 2 | 54 | 68 | ||
XM_025149502.1 | XP_025005270.1 | 1 - 2 | 4450 | 68 | ||
GenBank mRNA | AF082329.1 | - | 1 - 2 | 48 | 68 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
