Assay Details
| Gene Symbol: TUBB1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_205445.1 | NP_990776.1 | 3 - 4 | 351 | 67 | |
| XM_015296344.2 | XP_015151830.1 | 3 - 4 | 67 | |||
| GenBank mRNA | J02828.1 | - | 3 - 4 | 351 | 67 | |
More Information
Set Membership: |
|

TUBB1
Genomic Map